Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Chlorophenyl)biguanide hydrochloride, 97%

$200.11 - $200.11

Chemical Identifiers

Chemical Name or Material 1-(3-Chlorophenyl)biguanide hydrochloride
CAS 2113-05-5
Molecular Formula C8H10ClN5·HCl
Formula Weight 248.12
MDL Number MFCD00053019
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2035506

  SDS
alfa aesar™
B2035506
5g Each for $200.11
Description & Specifications

Specifications

Formula Weight 248.12
Chemical Name or Material 1-(3-Chlorophenyl)biguanide hydrochloride
Assay 97%
Molecular Formula C8H10ClN5·HCl
MDL Number MFCD00053019
CAS 2113-05-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 191° to 194°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.