Please login to your online account to display your discounted pricing

1-[3-chloro-5-(trifluoromethyl)-2-pyridyl]piperazine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]piperazine
CAS 132834-59-4
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H11ClF3N3
Formula Weight 265.66
MDL Number MFCD00173934
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW04036CB

 
fisher scientific company
SEW04036CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 265.66
Quantity 250mg
Assay 97%
CAS Min % 97.0
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10H11ClF3N3
MDL Number MFCD00173934
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-[3-Chloro-5-(trifluoromethyl)-2-pyridyl]piperazine
CAS 132834-59-4
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.