Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Chloro-4-fluorophenyl)piperidine, 98%

Chemical Identifiers

Chemical Name or Material 1-(3-Chloro-4-fluorophenyl)piperidine
CAS 1033201-89-6
Formula Weight 213.68
MDL Number MFCD10699654
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6135206

 
alfa aesar™
H6135206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6135214

 
alfa aesar™
H6135214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.68
Quantity 5g
Physical Form Solid
Assay 98%
MDL Number MFCD10699654
CAS 1033201-89-6
Chemical Name or Material 1-(3-Chloro-4-fluorophenyl)piperidine