Please login to your online account to display your discounted pricing

1-(3-Bromopropoxy)-3-chlorobenzene, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(3-Bromopropoxy)-3-chlorobenzene
CAS 37142-46-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C9H10BrClO
Linear Formula ClC6H4O(CH2)3Br
Formula Weight 249.53
MDL Number MFCD00596672
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379230050

  SDS
thermo fisher scientific
AC379230050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC379230250

  SDS
thermo fisher scientific
AC379230250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 249.53
Quantity 5g
Color Clear colorless
CAS Min % 95.0
CAS Max % 100.0
Linear Formula ClC6H4O(CH2)3Br
MDL Number MFCD00596672
CAS 37142-46-4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5570 to 1.5590 (20°C, 589nm)
Chemical Name or Material 1-(3-Bromopropoxy)-3-chlorobenzene
Assay Percent Range 96%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C9H10BrClO
Infrared Spectrum Authentic
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.