missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(3-Bromopropoxy)-3-chlorobenzene, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC379230250

 View more versions of this product

Catalog No. AC379230250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

249.53
≥95%
Clear colorless
100.0
ClC6H4O(CH2)3Br
Glass bottle
96%
MFCD00596672
Liquid
25g
95.0
1.5570 to 1.5590 (20°C, 589nm)
37142-46-4
Authentic
1-(3-Bromopropoxy)-3-chlorobenzene
C9H10BrClO

Chemical Identifiers

37142-46-4
100.0
C9H10BrClO
249.53
1-(3-Bromopropoxy)-3-chlorobenzene
95.0
96%
ClC6H4O(CH2)3Br
MFCD00596672
≥95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.