missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(3-Bromo-2-fluorophenyl)ethanol, 98%

$25.91 - $305.34

Chemical Identifiers

CAS 1221715-80-5
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.053
MDL Number MFCD16140247
InChI Key QFVPIDFXSBXICP-UHFFFAOYSA-N
Synonym 1-3-bromo-2-fluorophenyl ethanol, 1-3-bromo-2-fluoro-phenyl ethanol, 1-3-bromo-2-fluorophenyl ethan-1-ol, 3-bromo-2-fluoro-alpha-methylbenzenemethanol
PubChem CID 53484284
IUPAC Name 1-(3-bromo-2-fluorophenyl)ethanol
SMILES CC(C1=C(C(=CC=C1)Br)F)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64736MD View Documents Alfa Aesar™
H64736MD
250mg Each for $25.91
 
AAH6473603 View Documents Alfa Aesar™
H6473603
1g Each for $76.65
 
AAH6473606 View Documents Alfa Aesar™
H6473606
5g Each for $305.34
 
Specifications

Chemical Identifiers

1221715-80-5
219.053
QFVPIDFXSBXICP-UHFFFAOYSA-N
53484284
CC(C1=C(C(=CC=C1)Br)F)O
C8H8BrFO
MFCD16140247
1-3-bromo-2-fluorophenyl ethanol, 1-3-bromo-2-fluoro-phenyl ethanol, 1-3-bromo-2-fluorophenyl ethan-1-ol, 3-bromo-2-fluoro-alpha-methylbenzenemethanol
1-(3-bromo-2-fluorophenyl)ethanol

Specifications

250mg
C8H8BrFO
1-3-bromo-2-fluorophenyl ethanol, 1-3-bromo-2-fluoro-phenyl ethanol, 1-3-bromo-2-fluorophenyl ethan-1-ol, 3-bromo-2-fluoro-alpha-methylbenzenemethanol
CC(C1=C(C(=CC=C1)Br)F)O
219.053
219.05
1-(3-Bromo-2-fluorophenyl)ethanol
1221715-80-5
MFCD16140247
QFVPIDFXSBXICP-UHFFFAOYSA-N
1-(3-bromo-2-fluorophenyl)ethanol
53484284
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning