missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Bis[tris(hydroxymethyl)amino]propane, 99%, Acros Organics™

Manufacturer:  Acros Organics AC202640250

 View more versions of this product

Catalog No. AC202640250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1,3-Bis[tris(hydroxymethyl)amino]propane
Authentic
White
25g
98.5
99%
CH2{CH2NHC(CH2OH)3]2
04,III,859
Solubility in water: soluble in cold water
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
282.33
CHEBI:40947
Crystalline Powder
46.33 to 47.2% C, 9.82 to 10.02% N
Glass bottle
164.0°C to 165.0°C
64431-96-5
100.0
C11H26N2O6
MFCD00004689
1,3-bis tris hydroxymethyl methylamino propane
HHKZCCWKTZRCCL-UHFFFAOYSA-N
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
125132
282.33
99%

Chemical Identifiers

64431-96-5
282.33
HHKZCCWKTZRCCL-UHFFFAOYSA-N
125132
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
C11H26N2O6
MFCD00004689
1,3-bis tris hydroxymethyl methylamino propane
CHEBI:40947
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
SDS Documents