Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis[(trimethylsilyl)ethynyl]benzene, 96%

$54.39 - $180.81

Chemical Identifiers

Chemical Name or Material 1,3-Bis[(trimethylsilyl)ethynyl]benzene
CAS 38170-80-8
Molecular Formula C16H22Si2
Formula Weight 270.53
MDL Number MFCD00078305
Beilstein 2845239
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1064003

  SDS
alfa aesar™
L1064003
1g Each for $54.39

AAL1064006

  SDS
alfa aesar™
L1064006
5g Each for $180.81
Description & Specifications

Specifications

Formula Weight 270.53
CAS 38170-80-8
Chemical Name or Material 1,3-Bis[(trimethylsilyl)ethynyl]benzene
Assay 96%
MDL Number MFCD00078305
Molecular Formula C16H22Si2
Beilstein 2845239
Quantity 1g
Melting Point 57° to 59°C