Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(trimethylsilyl)benzene

$85.43 - $329.99

Chemical Identifiers

Chemical Name or Material 1,3-Bis(trimethylsilyl)benzene
CAS 2060-89-1
Formula Weight 222.48
MDL Number MFCD00015591
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1754203

  SDS
alfa aesar™
1754203
1g Each for $85.43

AA1754206

  SDS
alfa aesar™
1754206
5g Each for $329.99
Description & Specifications

Specifications

Formula Weight 222.48
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Health Hazard 1 Warning
MDL Number MFCD00015591
Boiling Point 120°C (25mmHg)
Refractive Index 1.482
CAS 2060-89-1
Chemical Name or Material 1,3-Bis(trimethylsilyl)benzene
Quantity 1g
Flash Point >60° (140°F)
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.