missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(trifluoromethyl)benzene, 98+%

Manufacturer:  Alfa Aesar™ A1136018

 View more versions of this product

Catalog No. AAA1136018


 

Description

Description

Applications
1,3-Bis(trifluoromethyl)benzene undergoes regioselective metalation and subsequent carboxylation at position 2 to give 2,6-bis(trifluoromethyl)benzoic acid. a convenient, selective synthesis of bis[2,4-bis(trifluoromethyl)phenyl]phosphane derivatives with 1,3-bis(trifluoromethyl)benzene as starting material.

Solubility
Insoluble in water. Soluble in alcohol, ether, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

1,3-Bis (trifluoromethyl)benzene
≥98%
MFCD00000392
2052589
-35°C
SJBBXFLOLUTGCW-UHFFFAOYSA-N
26°C (78°F)
1,3-bis(trifluoromethyl)benzene
214.11
214.11
402-31-3
C8H4F6
UN1993
alpha,alpha,alpha,alpha',alpha',alpha'-Hexafluoro-m-xylene
115°C to 117°C
50g
C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Aromatic
223106

Chemical Identifiers

402-31-3
214.11
SJBBXFLOLUTGCW-UHFFFAOYSA-N
223106
C8H4F6
MFCD00000392
alpha,alpha,alpha,alpha',alpha',alpha'-Hexafluoro-m-xylene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P260-P280g-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents