Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(phenylphosphino)propane

$89.80 - $89.80

Chemical Identifiers

Chemical Name or Material 1,3-Bis(phenylphosphino)propane
CAS 28240-66-6
Formula Weight 260.26
MDL Number MFCD00014079
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3358902

  SDS
alfa aesar™
3358902
0.5g Each for $89.80

AA3358904

  SDS
alfa aesar™
3358904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid, Ampouled Under Argon
Formula Weight 260.26
Chemical Name or Material 1,3-Bis(phenylphosphino)propane
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 0.5g
MDL Number MFCD00014079
Sensitivity Air Sensitive
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 160° to 165°C (1mmHg)
CAS 28240-66-6
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.