Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(methylthio)propane, 97%

$116.92 - $116.92

Chemical Identifiers

Chemical Name or Material 1,3-Bis(methylthio)propane
CAS 24949-35-7
Molecular Formula CH3S(CH2)3SCH3
Formula Weight 136.28
MDL Number MFCD00039811
Beilstein 1732333
Synonym 1,3-Bis(methylthio)propane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2239703

  SDS
alfa aesar™
B2239703
1g Each for $116.92
Description & Specifications

Specifications

Formula Weight 136.28
Synonym 1,3-Bis(methylthio)propane
Flash Point >100°C (212°F)
Molecular Formula CH3S(CH2)3SCH3
Boiling Point 88° to 90°C (12mmHg)
Quantity 1g
Beilstein 1732333
Odor Stench
MDL Number MFCD00039811
CAS 24949-35-7
Chemical Name or Material 1,3-Bis(methylthio)propane
Assay 97%