Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(methylthio)-2-propanol, 97%

$38.95 - $130.84

Chemical Identifiers

Chemical Name or Material 1,3-Bis(methylthio)-2-propanol
CAS 31805-83-1
Molecular Formula CH3SCH2CH(OH)CH2SCH3
Formula Weight 152.29
MDL Number MFCD00041435
Beilstein 1734314
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2189303

 
alfa aesar™
B2189303
1g Each for $38.95

AAB2189306

 
alfa aesar™
B2189306
5g Each for $130.84
Description & Specifications

Specifications

Formula Weight 152.29
Chemical Name or Material 1,3-Bis(methylthio)-2-propanol
Assay 97%
Molecular Formula CH3SCH2CH(OH)CH2SCH3
MDL Number MFCD00041435
CAS 31805-83-1
Beilstein 1734314
Boiling Point 112° to 114°C (5mmHg)
Quantity 1g