Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(hydroxymethyl)urea, tech. 90%

$54.19 - $538.74

Chemical Identifiers

Chemical Name or Material 1,3-Bis(hydroxymethyl)urea
CAS 140-95-4
Purity Grade Technical Grade
Molecular Formula C3H8N2O3
Linear Formula (HOCH2NH)2C=O
Formula Weight 120.11
MDL Number MFCD00014414
Synonym Dimethylolurea; DMU
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA160600E

 
alfa aesar™
A160600E
2500g Each for $538.74

AAA1606022

 
alfa aesar™
A1606022
100g Each for $54.19

AAA1606036

 
alfa aesar™
A1606036
500g Each for $141.30
Description & Specifications

Specifications

Formula Weight 120.11
Chemical Name or Material 1,3-Bis(hydroxymethyl)urea
TSCA TSCA
Purity Grade Technical Grade
Assay 90%
Molecular Formula C3H8N2O3
Linear Formula (HOCH2NH)2C=O
MDL Number MFCD00014414
CAS 140-95-4
Synonym Dimethylolurea; DMU
Melting Point 124° to 128°C
pH 7 to 8
Quantity 2500g
Odor Odorless