missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Bis(hexafluoro-alpha-hydroxyisopropyl)benzene, TCI America™

$137.50 - $403.34

Chemical Identifiers

CAS 802-93-7
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.159
MDL Number MFCD00042090
InChI Key PGUIOHNOYADLMU-UHFFFAOYSA-N
Synonym 1,3-bis 2-hydroxyhexafluoroisopropyl benzene, 1,3-bis hexafluoro-2-hydroxyisopropyl benzene, 1,3-bis alpha-hydroxyhexafluoroisopropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,3-benzenedimethanol, a,a,a',a'-tetrakis trifluoromethyl-1,3-benzenedimethanol, 1,3-benzenedimethanol, .alpha.,.alpha.,.alpha.',.alpha.'-tetrakis trifluoromethyl, 1,3-bis hexafluoro-alpha-hydroxyisopropyl benzene, 1,1,1,3,3,3-hexafluoro-2-3-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,3-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, 1,3 hfab
PubChem CID 69933
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES C1=CC(=CC(=C1)C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14035G View Documents TCI America
B14035G
5g Each for $137.50
 
B140325G View Documents TCI America
B140325G
25g Each for $403.34
 
Specifications

Chemical Identifiers

802-93-7
410.159
PGUIOHNOYADLMU-UHFFFAOYSA-N
69933
C1=CC(=CC(=C1)C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O
C12H6F12O2
MFCD00042090
1,3-bis 2-hydroxyhexafluoroisopropyl benzene, 1,3-bis hexafluoro-2-hydroxyisopropyl benzene, 1,3-bis alpha-hydroxyhexafluoroisopropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,3-benzenedimethanol, a,a,a',a'-tetrakis trifluoromethyl-1,3-benzenedimethanol, 1,3-benzenedimethanol, .alpha.,.alpha.,.alpha.',.alpha.'-tetrakis trifluoromethyl, 1,3-bis hexafluoro-alpha-hydroxyisopropyl benzene, 1,1,1,3,3,3-hexafluoro-2-3-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,3-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, 1,3 hfab
1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol

Specifications

100°C
5g
C12H6F12O2
1,3-bis 2-hydroxyhexafluoroisopropyl benzene, 1,3-bis hexafluoro-2-hydroxyisopropyl benzene, 1,3-bis alpha-hydroxyhexafluoroisopropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,3-benzenedimethanol, a,a,a',a'-tetrakis trifluoromethyl-1,3-benzenedimethanol, 1,3-benzenedimethanol, .alpha.,.alpha.,.alpha.',.alpha.'-tetrakis trifluoromethyl, 1,3-bis hexafluoro-alpha-hydroxyisopropyl benzene, 1,1,1,3,3,3-hexafluoro-2-3-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,3-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, 1,3 hfab
C1=CC(=CC(=C1)C(C(F)(F)F)(C(F)(F)F)O)C(C(F)(F)F)(C(F)(F)F)O
410.159
410.16
≥98.0% (GC)
Yellow
802-93-7
MFCD00042090
PGUIOHNOYADLMU-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoro-2-[3-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
69933
Clear Liquid at 20°C
1,3-Bis(hexafluoro-alpha-hydroxyisopropyl)benzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4346

TSCA : Yes