missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 291590050
Specifications
[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride | |
Authentic | |
Undesignated | |
5g | |
98.5 | |
99% | |
MFCD00015318 | |
Solubility in water: insoluble | |
C1=CC=C(C=C1)P(CCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni] | |
542.04 | |
542.04 | |
99% |
98.5% min. | |
Glass bottle | |
213.0°C | |
15629-92-2 | |
100.0 | |
C27H26Cl2NiP2 | |
06,50; 15,122 | |
BILYTLYIOVUVSU-UHFFFAOYSA-N | |
3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride | |
131675641 | |
Powder or Crystals |
Chemical Identifiers
15629-92-2 | |
542.04 | |
BILYTLYIOVUVSU-UHFFFAOYSA-N | |
3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride |
C27H26Cl2NiP2 | |
MFCD00015318 | |
131675641 | |
C1=CC=C(C=C1)P(CCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl.Cl.[Ni] |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON
GHS Signal Word: Danger