missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-bis(dicyclohexylphosphino)propane, 95%, ACROS Organics™

Chemical Identifiers

CAS 103099-52-1
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C27H50P2
Formula Weight 436.64
MDL Number MFCD00799482
Chemical Name or Material 1, 3-Bis(dicyclohexylphosphino)propane
Assay ≥94.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC381120010 SDS Thermo Fisher Scientific
AC381120010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC381122500 SDS Thermo Fisher Scientific
AC381122500
Glass Bottle in Plastic Container 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

103099-52-1
100.0
C27H50P2
MFCD00799482
≥94.0%
94.0
95%
436.64
1, 3-Bis(dicyclohexylphosphino)propane

Specifications

Paste
1g
94.0
Authentic
MFCD00799482
95%
1, 3-Bis(dicyclohexylphosphino)propane
436.64
Clear colorless
100.0
C27H50P2
103099-52-1
Glass bottle
≥94.0%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning