Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(dichloromethyl)-1,1,3,3-tetramethyldisiloxane, 94%

$298.62 - $298.62

Chemical Identifiers

Chemical Name or Material 1,3-Bis(dichloromethyl)-1,1,3,3-tetramethyldisiloxane
CAS 2943-70-6
Molecular Formula C6H14Cl4OSi2
Formula Weight 300.16
MDL Number MFCD00040856
Beilstein 1762720
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0585822

 
alfa aesar™
L0585822
100g Each for $298.62
Description & Specifications

Specifications

Formula Weight 300.16
Chemical Name or Material 1,3-Bis(dichloromethyl)-1,1,3,3-tetramethyldisiloxane
Assay 94%
Molecular Formula C6H14Cl4OSi2
CAS 2943-70-6
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
MDL Number MFCD00040856
Beilstein 1762720
Boiling Point 149° to 150°C (40mmHg)
Quantity 100g
Flash Point 65°C (149°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.