Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(chloromethyl)tetramethyldisiloxane, 97%

$44.15 - $164.24

Chemical Identifiers

Chemical Name or Material 1,3-Bis(chloromethyl)tetramethyldisiloxane
CAS 2362-10-9
Molecular Formula C6H16Cl2OSi2
Formula Weight 231.27
MDL Number MFCD00000877
Beilstein 506631
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0443609

  SDS
alfa aesar™
L0443609
10g Each for $44.15

AAL0443618

  SDS
alfa aesar™
L0443618
50g Each for $164.24
Description & Specifications

Specifications

Formula Weight 231.27
MDL Number MFCD00000877
Chemical Name or Material 1,3-Bis(chloromethyl)tetramethyldisiloxane
CAS 2362-10-9
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Assay 97%
Molecular Formula C6H16Cl2OSi2
Quantity 10g
Health Hazard 1 Warning
Flash Point 73°C (163°F)
TSCA TSCA
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 506631
Boiling Point 92° to 93°C (20mmHg)

Applications
1,3-Bis(chloromethyl)tetramethyldisiloxane is a useful bifunctional silylating agent.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.