missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Bis(4-methoxyphenyl)-1,3-propanedione 98.0+%, TCI America™
$296.97 - $892.54
Chemical Identifiers
CAS | 18362-51-1 |
---|---|
Molecular Formula | C17H16O4 |
Molecular Weight (g/mol) | 284.31 |
MDL Number | MFCD00025817 |
InChI Key | GNMDORSUZRRMFS-UHFFFAOYSA-N |
PubChem CID | 87596 |
IUPAC Name | 1,3-bis(4-methoxyphenyl)propane-1,3-dione |
SMILES | COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(OC)C=C1 |
Chemical Identifiers
18362-51-1 | |
284.31 | |
GNMDORSUZRRMFS-UHFFFAOYSA-N | |
1,3-bis(4-methoxyphenyl)propane-1,3-dione |
C17H16O4 | |
MFCD00025817 | |
87596 | |
COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(OC)C=C1 |
Specifications
18362-51-1 | |
C17H16O4 | |
5 g | |
COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(OC)C=C1 | |
284.31 | |
284.31 | |
Crystalline Powder |
116°C | |
MFCD00025817 | |
GNMDORSUZRRMFS-UHFFFAOYSA-N | |
1,3-bis(4-methoxyphenyl)propane-1,3-dione | |
87596 | |
≥98.0% (GC) | |
1,3-Bis(4-methoxyphenyl)-1,3-propanedione |
Safety and Handling
TSCA : Yes