Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(4-methoxyphenyl)-1,1,3,3-tetramethyldisiloxane, 97%

$51.88 - $166.24

Chemical Identifiers

Chemical Name or Material 1,3-Bis(4-methoxyphenyl)-1,1,3,3-tetramethyldisiloxane
CAS 122571-17-9
Molecular Formula C18H26O3Si2
Formula Weight 346.57
MDL Number MFCD09266156
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2847703

  SDS
alfa aesar™
H2847703
1g Each for $51.88

AAH2847706

  SDS
alfa aesar™
H2847706
5g Each for $166.24
Description & Specifications

Specifications

Formula Weight 346.57
Chemical Name or Material 1,3-Bis(4-methoxyphenyl)-1,1,3,3-tetramethyldisiloxane
Assay 97%
Molecular Formula C18H26O3Si2
MDL Number MFCD09266156
CAS 122571-17-9
Quantity 1g