Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate, 98%

$75.42 - $75.42

Chemical Identifiers

Chemical Name or Material 1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate
CAS 915967-82-7
Formula Weight 999.55
MDL Number MFCD11870901
Synonym Piperaquine tetraphosphate tetrahydrate; 7-Chloro-4-[4-(3-[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propyl)-1-piperazinyl]quinoline tetraphosphate tetrahydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6651314

 
alfa aesar™
H6651314
25g Each for $75.42

AAH6651322

 
alfa aesar™
H6651322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 999.55
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate
Physical Form Powder
MDL Number MFCD11870901
CAS 915967-82-7
Color Off-white to white
Quantity 25g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Piperaquine tetraphosphate tetrahydrate; 7-Chloro-4-[4-(3-[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propyl)-1-piperazinyl]quinoline tetraphosphate tetrahydrate
Sensitivity Hygroscopic
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.