missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Bis(3-aminophenoxy)-5-chlorobenzene Dihydrochloride, TCI America™

$345.28 - $1,086.27

Chemical Identifiers

CAS 99503-73-8
Molecular Formula C18H17Cl3N2O2
Molecular Weight (g/mol) 399.696
InChI Key YZUFDVOYHZOIGU-UHFFFAOYSA-N
PubChem CID 44630035
IUPAC Name 3-[3-(3-aminophenoxy)-5-chlorophenoxy]aniline;dihydrochloride
SMILES C1=CC(=CC(=C1)OC2=CC(=CC(=C2)Cl)OC3=CC=CC(=C3)N)N.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B35501G View Documents TCI America
B35501G
1g Each for $345.28
 
B35505G View Documents TCI America
B35505G
5g Each for $1,086.27
 
Specifications

Chemical Identifiers

99503-73-8
399.696
44630035
C1=CC(=CC(=C1)OC2=CC(=CC(=C2)Cl)OC3=CC=CC(=C3)N)N.Cl.Cl
C18H17Cl3N2O2
YZUFDVOYHZOIGU-UHFFFAOYSA-N
3-[3-(3-aminophenoxy)-5-chlorophenoxy]aniline;dihydrochloride

Specifications

White
99503-73-8
YZUFDVOYHZOIGU-UHFFFAOYSA-N
3-[3-(3-aminophenoxy)-5-chlorophenoxy]aniline;dihydrochloride
44630035
Crystal-Powder at 20°C
1,3-Bis(3-aminophenoxy)-5-chlorobenzene Dihydrochloride
1g
C18H17Cl3N2O2
C1=CC(=CC(=C1)OC2=CC(=CC(=C2)Cl)OC3=CC=CC(=C3)N)N.Cl.Cl
399.696
399.70
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No