Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene, 96%

$88.15 - $302.86

Chemical Identifiers

Chemical Name or Material 1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
CAS 802-93-7
Molecular Formula C12H6F12O2
Formula Weight 410.16
MDL Number MFCD00042090
Beilstein 2024879
Synonym alpha,alpha,alpha',alpha'-Tetrakis(trifluoromethyl)-1,3-benzenedimethanol; 1,3-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1689406

  SDS
alfa aesar™
L1689406
5g Each for $88.15

AAL1689414

  SDS
alfa aesar™
L1689414
25g Each for $302.86
Description & Specifications

Specifications

Formula Weight 410.16
MDL Number MFCD00042090
Assay 96%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene
CAS 802-93-7
Molecular Formula C12H6F12O2
Synonym alpha,alpha,alpha',alpha'-Tetrakis(trifluoromethyl)-1,3-benzenedimethanol; 1,3-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)benzene
Flash Point 97°C (206°F)
Beilstein 2024879
TSCA TSCA
Boiling Point 99° to 100°C (20mmHg)
Melting Point 9° to 10°C
Quantity 5g

Applications
1,3-Bis(2-hydroxyhexafluoroisopropyl)benzene is used to determine the specific details of electrical magnitudes.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.