missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Bis(2-hydroxyethylsulfonyl)-2-propanol, TCI America™

$90.14 - $637.09

Chemical Identifiers

CAS 67006-34-2
Molecular Formula C7H16O7S2
Molecular Weight (g/mol) 276.318
InChI Key CVFXDAXIZHJZAH-UHFFFAOYSA-N
PubChem CID 11437335
IUPAC Name 1,3-bis(2-hydroxyethylsulfonyl)propan-2-ol
SMILES C(CS(=O)(=O)CC(CS(=O)(=O)CCO)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B269925G View Documents TCI America
B269925G
25g Each for $90.14
 
B2699500G View Documents TCI America
B2699500G
500g Each for $637.09
 
Specifications

Chemical Identifiers

67006-34-2
276.318
11437335
C(CS(=O)(=O)CC(CS(=O)(=O)CCO)O)O
C7H16O7S2
CVFXDAXIZHJZAH-UHFFFAOYSA-N
1,3-bis(2-hydroxyethylsulfonyl)propan-2-ol

Specifications

White-Yellow
67006-34-2
CVFXDAXIZHJZAH-UHFFFAOYSA-N
1,3-bis(2-hydroxyethylsulfonyl)propan-2-ol
11437335
Crystal-Powder at 20°C
1,3-Bis(2-hydroxyethylsulfonyl)-2-propanol
25g
C7H16O7S2
C(CS(=O)(=O)CC(CS(=O)(=O)CCO)O)O
276.318
276.32
≥93.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No