missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Bis(2-hydroxyethoxy)benzene 98.0+%, TCI America™
Supplier: TCI America B1625500G
Specifications
1,3-Bis(2-hydroxyethoxy)benzene | |
90°C | |
234°C | |
MFCD00016566 | |
Resorcinol Bis(beta-hydroxyethyl) Ether | |
OCCOC1=CC(OCCO)=CC=C1 | |
198.22 | |
198.22 | |
Crystalline Powder |
102-40-9 | |
White | |
C10H14O4 | |
500 g | |
IAXFZZHBFXRZMT-UHFFFAOYSA-N | |
2-[3-(2-hydroxyethoxy)phenoxy]ethan-1-ol | |
66885 | |
≥98.0% (GC) |
Chemical Identifiers
102-40-9 | |
198.22 | |
IAXFZZHBFXRZMT-UHFFFAOYSA-N | |
66885 | |
OCCOC1=CC(OCCO)=CC=C1 |
C10H14O4 | |
MFCD00016566 | |
Resorcinol Bis(beta-hydroxyethyl) Ether | |
2-[3-(2-hydroxyethoxy)phenoxy]ethan-1-ol |
Safety and Handling
EINECSNumber : (3)-3864
TSCA : Yes