Please login to your online account to display your discounted pricing

1,3-Bis(2,6-diisopropylphenyl)imidazolidinium tetrafluoroborate, +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium tetrafluoroborate
CAS 282109-83-5
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Molecular Formula C27H39BF4N2
Formula Weight 478.42
MDL Number MFCD04971187
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382420050

  SDS
thermo fisher scientific
AC382420050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC382420010

  SDS
fisher scientific acros
AC382420010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 478.42
Quantity 5g
Color White
CAS Min % 97.0
CAS Max % 100.0
Chemical Name or Material 1, 3-Bis(2, 6-diisopropylphenyl)imidazolidinium tetrafluoroborate
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 97+%
Molecular Formula C27H39BF4N2
MDL Number MFCD04971187
CAS 282109-83-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Elemental Analysis 97.0% min.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.