Learn More
1,3-bis(2,4,6-trimethylphenyl)imidazolidinium chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 378330010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1,3-Bis(2,4,6-trimethylphenyl)imidazolidinium chloride | |
96.0 | |
280°C to 286°C | |
Authentic | |
Glass bottle | |
MFCD09039279 | |
1,3-dimesitylimidazolidine hydrochloride, 1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride, 1,3-dimesitylimidazolidine, chloride salt, 1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride, 1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride | |
[Cl-].CC1=CC(C)=C(N2CC[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1 | |
342.91 | |
97% |
173035-10-4 | |
100.0 | |
Beige to Brown-Pink | |
96% min. (HPLC) | |
C21H27ClN2 | |
1 g | |
COGMCBFILULEOS-UHFFFAOYSA-M | |
1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λâµ-imidazol-1-ylium chloride | |
342.91 | |
Powder |
Chemical Identifiers
173035-10-4 | |
342.91 | |
COGMCBFILULEOS-UHFFFAOYSA-M | |
1,3-bis(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λâµ-imidazol-1-ylium chloride |
C21H27ClN2 | |
MFCD09039279 | |
1,3-dimesitylimidazolidine hydrochloride, 1,3-bis 2,4,6-trimethylphenyl-imidazolidinium-chloride, 1,3-dimesitylimidazolidine, chloride salt, 1,3-bis-2,4,6-trimethyl-phenyl-imidazolidin-1-ium chloride, 1,3-bis 2,4,6-trimethylphenyl imidazolidine hydrochloride | |
[Cl-].CC1=CC(C)=C(N2CC[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
RUO – Research Use Only