missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate

Manufacturer:  Alfa Aesar™ H3633006

 View more versions of this product

Catalog No. AAH3633006



Specifications

Specifications

1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate
280°C to 286°C
286014-42-4
MFCD04973311
KVWCCJYLKCSVME-UHFFFAOYSA-N
1,3-bis(1-adamantyl)imidazol-1-ium;tetrafluoroborate
16218105
Powder
Brown to White
5g
C23H33BF4N2
1,3-bis 1-adamantyl imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazolium tetrafluoroborate, 1,3-di adamantan-1-yl-1h-imidazol-3-ium tetrafluoroborate, 1,3-bis adamantan-1-yl-1??-imidazol-1-ylium tetrafluoroborate, iad.hbf4, acmc-209h3n, 1,3-diadamantyl-imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazoliumtetrafluoroborate, 1,3-bis 1-adamantyl imidazolium tetrafluoroborate, 1,3-bis tricyclo 3.3.1.13,7 dec-1-yl-1h-imidazolium tetrafluoroborate
[B-](F)(F)(F)F.C1C2CC3CC1CC(C2)(C3)N4C=C[N+](=C4)C56CC7CC(C5)CC(C7)C6
424.335
424.33
96%

Chemical Identifiers

286014-42-4
424.335
KVWCCJYLKCSVME-UHFFFAOYSA-N
16218105
[B-](F)(F)(F)F.C1C2CC3CC1CC(C2)(C3)N4C=C[N+](=C4)C56CC7CC(C5)CC(C7)C6
C23H33BF4N2
MFCD04973311
1,3-bis 1-adamantyl imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazolium tetrafluoroborate, 1,3-di adamantan-1-yl-1h-imidazol-3-ium tetrafluoroborate, 1,3-bis adamantan-1-yl-1??-imidazol-1-ylium tetrafluoroborate, iad.hbf4, acmc-209h3n, 1,3-diadamantyl-imidazolium tetrafluoroborate, 1,3-di 1-adamantyl imidazoliumtetrafluoroborate, 1,3-bis 1-adamantyl imidazolium tetrafluoroborate, 1,3-bis tricyclo 3.3.1.13,7 dec-1-yl-1h-imidazolium tetrafluoroborate
1,3-bis(1-adamantyl)imidazol-1-ium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents