missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Bis(1-adamantyl)benzimidazolium chloride, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC435070010

 View more versions of this product

Catalog No. AC435070010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1, 3-Bis(1-Adamantyl)benzimidazolium chloride
Authentic
Gray to White
1g
94.0
94% min. (HPLC)
1,3-bis 1-adamanthyl benzimidazolium chloride, 1,3-bis 1-adamantyl benzimidazolium chloride, 1,3-bis 1-adamantyl-benzimidazolium chloride, 1,3-bis 3r,5s,7s-adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride, 1,3-bis adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride, 1,3-bis adamantan-1-yl-2h-1,3-benzodiazol-2-ylium chloride, 1,3-di adamantan-1-yl-1h-benzo d imidazol-3-ium chloride
C1C2CC3CC1CC(C2)(C3)N4C=[N+](C5=CC=CC=C54)C67CC8CC(C6)CC(C8)C7.[Cl-]
423.05
423.05
95%
72.06 to 81.26% C, 6.22 to 7.02% N
Glass Bottle
283.9°C to 285.6°C
852634-41-4
100.0
C27H35N2·Cl
GMYUUHXEUIPSKA-UHFFFAOYSA-M
1,3-bis(1-adamantyl)benzimidazol-3-ium;chloride
16218257
Powder

Chemical Identifiers

852634-41-4
423.05
1,3-bis 1-adamanthyl benzimidazolium chloride, 1,3-bis 1-adamantyl benzimidazolium chloride, 1,3-bis 1-adamantyl-benzimidazolium chloride, 1,3-bis 3r,5s,7s-adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride, 1,3-bis adamantan-1-yl-1??,3-benzodiazol-1-ylium chloride, 1,3-bis adamantan-1-yl-2h-1,3-benzodiazol-2-ylium chloride, 1,3-di adamantan-1-yl-1h-benzo d imidazol-3-ium chloride
1,3-bis(1-adamantyl)benzimidazol-3-ium;chloride
C27H35N2·Cl
GMYUUHXEUIPSKA-UHFFFAOYSA-M
16218257
C1C2CC3CC1CC(C2)(C3)N4C=[N+](C5=CC=CC=C54)C67CC8CC(C6)CC(C8)C7.[Cl-]
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents