Please login to your online account to display your discounted pricing

1,3-Bis(1-adamantyl)benzimidazolium chloride, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Bis(1-Adamantyl)benzimidazolium chloride
CAS 852634-41-4
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C27H35ClN2
Formula Weight 423.05
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC435070010

 
thermo fisher scientific
AC435070010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC435075000

 
thermo fisher scientific
AC435075000
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥94.0%
Formula Weight 423.05
Quantity 1g
Color White to tan to gray
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 1, 3-Bis(1-Adamantyl)benzimidazolium chloride
Assay Percent Range 95%
Molecular Formula C27H35ClN2
CAS 852634-41-4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Elemental Analysis 72.06 to 81.26% C, 6.22 to 7.02% N
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.