Learn More
Thermo Scientific Chemicals 1,3-Benzothiazole-6-sulfonyl chloride, ≥95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC05903CB
Specifications
1, 3-Benzothiazole-6-sulfonyl chloride | |
94.5 | |
95% | |
C7H4ClNO2S2 | |
250 mg | |
XQOLJTWXFUSVOR-UHFFFAOYSA-N | |
1,3-benzothiazole-6-sulfonyl chloride | |
2795170 | |
95% |
181124-40-3 | |
100.0 | |
Amber Glass Bottle | |
MFCD03086100 | |
benzo d thiazole-6-sulfonyl chloride, 1,3-benzothiazole-6-sulfonylchloride, 6-benzothiazolesulfonyl chloride, 1,3-benzothiazole-6-sulphonyl chloride, 6-chlorosulfonylbenzothiazole, buttpark 121\04-40, 1,3-benzothiazole, benzothiazol-6-ylchlorosulfone, 6-benzothiazolesulfonylchloride, benzothiazole-6-sulphonyl chloride | |
C1=CC2=C(C=C1S(=O)(=O)Cl)SC=N2 | |
233.684 | |
233.7 |
Chemical Identifiers
181124-40-3 | |
233.684 | |
XQOLJTWXFUSVOR-UHFFFAOYSA-N | |
2795170 | |
C1=CC2=C(C=C1S(=O)(=O)Cl)SC=N2 |
C7H4ClNO2S2 | |
MFCD03086100 | |
benzo d thiazole-6-sulfonyl chloride, 1,3-benzothiazole-6-sulfonylchloride, 6-benzothiazolesulfonyl chloride, 1,3-benzothiazole-6-sulphonyl chloride, 6-chlorosulfonylbenzothiazole, buttpark 121\04-40, 1,3-benzothiazole, benzothiazol-6-ylchlorosulfone, 6-benzothiazolesulfonylchloride, benzothiazole-6-sulphonyl chloride | |
1,3-benzothiazole-6-sulfonyl chloride |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger