Please login to your online account to display your discounted pricing

1,3-Benzothiazole-6-carboxaldehyde, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Benzothiazole-6-carboxaldehyde
CAS 19989-67-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (HPLC)
Assay Percent Range 97%
Molecular Formula C8H5NOS
Formula Weight 163.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC431150010

  SDS
thermo fisher scientific
AC431150010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC431150050

  SDS
thermo fisher scientific
AC431150050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder or Low Melting Solid
Formula Weight 163.2
Assay ≥96.0% (HPLC)
Quantity 1g
Color White to yellow to brown
Melting Point 90° to 96°C
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
CAS 19989-67-4
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Chemical Name or Material 1, 3-Benzothiazole-6-carboxaldehyde
Molecular Formula C8H5NOS
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.