Please login to your online account to display your discounted pricing

1,3-Benzothiazol-6-ylmethylamine dihydrochloride, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC430100010

 View more versions of this product

Catalog No. AC430100010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 1, 3-Benzothiazol-6-ylmethylamine dihydrochloride
CAS 1375964-49-0
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Assay Percent Range 97%
Molecular Formula C8H8N22ClH
Formula Weight 237.15
Quantity 1g

Specifications

Chemical Name or Material 1, 3-Benzothiazol-6-ylmethylamine dihydrochloride
Assay Percent Range 97%
Molecular Formula C8H8N22ClH
Formula Weight 237.15
Accessories

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.