Please login to your online account to display your discounted pricing

1,3-Benzothiazol-6-ylmethylamine dihydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1, 3-Benzothiazol-6-ylmethylamine dihydrochloride
Assay Percent Range 97%
Molecular Formula C8H8N22ClH
Formula Weight 237.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC430100010

 
thermo fisher scientific
AC430100010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC430100050

 
thermo fisher scientific
AC430100050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 237.15
Quantity 1g
Chemical Name or Material 1, 3-Benzothiazol-6-ylmethylamine dihydrochloride
Assay Percent Range 97%
CAS 1375964-49-0
Molecular Formula C8H8N22ClH
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.