missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
1,3-Benzothiazol-2-ol, 97%, ACROS Organics™
Chemical Identifiers
CAS |
934-34-9 |
Molecular Formula |
C7H5NOS |
Molecular Weight (g/mol) |
151.183 |
MDL Number |
MFCD00022868 |
InChI Key |
YEDUAINPPJYDJZ-UHFFFAOYSA-N |
Synonym |
1,3-benzothiazol-2-ol, 2 3h-benzothiazolone, 2-benzothiazolinone, 2-benzothiazolol, 2-benzothiazolone, 2-hydroxybenzothiazole, 3h-benzothiazol-2-one, benzo d thiazol-2 3h-one, benzothiazolol, benzothiazolone |
PubChem CID |
13625 |
ChEBI |
CHEBI:115196 |
IUPAC Name |
3H-1,3-benzothiazol-2-one |
SMILES |
C1=CC=C2C(=C1)NC(=O)S2 |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
934-34-9 |
|
151.183 |
|
YEDUAINPPJYDJZ-UHFFFAOYSA-N |
|
13625 |
|
3H-1,3-benzothiazol-2-one |
|
C7H5NOS |
|
MFCD00022868 |
|
1,3-benzothiazol-2-ol, 2 3h-benzothiazolone, 2-benzothiazolinone, 2-benzothiazolol, 2-benzothiazolone, 2-hydroxybenzothiazole, 3h-benzothiazol-2-one, benzo d thiazol-2 3h-one, benzothiazolol, benzothiazolone |
|
CHEBI:115196 |
|
C1=CC=C2C(=C1)NC(=O)S2 |
Specifications
|
934-34-9 |
|
100.0 |
|
134°C to 140°C |
|
Authentic |
|
MFCD00022868 |
|
1g |
|
YEDUAINPPJYDJZ-UHFFFAOYSA-N |
|
3H-1,3-benzothiazol-2-one |
|
13625 |
|
151.19 |
|
≥94% |
|
94.5 |
|
White to Brown |
|
95% |
|
C7H5NOS |
|
Amber glass bottle |
|
1,3-benzothiazol-2-ol, 2 3h-benzothiazolone, 2-benzothiazolinone, 2-benzothiazolol, 2-benzothiazolone, 2-hydroxybenzothiazole, 3h-benzothiazol-2-one, benzo d thiazol-2 3h-one, benzothiazolol, benzothiazolone |
|
C1=CC=C2C(=C1)NC(=O)S2 |
|
151.183 |
|
CHEBI:115196 |
|
Crystalline Powder |
|
2-Benzothiazolol |
SDS
Product Certifications