missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Benzothiazol-2-ol, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Company AC40197DA

 View more versions of this product

Catalog No. AC40197DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Benzothiazolol
Amber glass bottle
134°C to 140°C
934-34-9
100.0
C7H5NOS
1,3-benzothiazol-2-ol, 2 3h-benzothiazolone, 2-benzothiazolinone, 2-benzothiazolol, 2-benzothiazolone, 2-hydroxybenzothiazole, 3h-benzothiazol-2-one, benzo d thiazol-2 3h-one, benzothiazolol, benzothiazolone
C1=CC=C2C(=C1)NC(=O)S2
151.183
CHEBI:115196
Crystalline Powder
Authentic
White to Brown
1g
94.5
95%
MFCD00022868
YEDUAINPPJYDJZ-UHFFFAOYSA-N
3H-1,3-benzothiazol-2-one
13625
151.19
≥94%

Chemical Identifiers

934-34-9
151.183
YEDUAINPPJYDJZ-UHFFFAOYSA-N
13625
3H-1,3-benzothiazol-2-one
C7H5NOS
MFCD00022868
1,3-benzothiazol-2-ol, 2 3h-benzothiazolone, 2-benzothiazolinone, 2-benzothiazolol, 2-benzothiazolone, 2-hydroxybenzothiazole, 3h-benzothiazol-2-one, benzo d thiazol-2 3h-one, benzothiazolol, benzothiazolone
CHEBI:115196
C1=CC=C2C(=C1)NC(=O)S2
SDS Documents