Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Benzodithiolylium tetrafluoroborate, 98%

Chemical Identifiers

Chemical Name or Material 1,3-Benzodithiolylium tetrafluoroborate
CAS 57842-27-0
Formula Weight 240.04
MDL Number MFCD00070613
Beilstein 4731453
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5540303

 
alfa aesar™
H5540303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5540306

 
alfa aesar™
H5540306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5540314

 
alfa aesar™
H5540314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.04
CAS 57842-27-0
Melting Point 150°C (dec.)
Color Gray to brown
Chemical Name or Material 1,3-Benzodithiolylium tetrafluoroborate
Physical Form Solid
Assay 98%
MDL Number MFCD00070613
Beilstein 4731453
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.