missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Benzodioxole, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 170220010

Catalog No. AC170220010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1, 3-Benzodioxole
274-09-9
100.0
172.0°C to 173.0°C
Authentic
Glass bottle
1.5380 to 1.5400
1kg
1.06
Solubility in water: 0.2% (25 c). Other solubilities: miscible with most common organic solvents
C1OC2=CC=CC=C2O1
122.12
CHEBI:38732
99%
99%
98.5
1.0600g/mL
55°C
98.5% min. (GC)
C7H6O2
MFCD00005818
19,2
1,2-methylenedioxybenzene, benzodioxole, benzo d 1,3 dioxole, 1,3-dioxaindan, 3,4-methylenedioxybenzene, 2h-1,3-benzodioxole, 1,2-methylenedioxy benzene, 1,3-dioxindan, methylenedioxybenzene, o-methylenedioxy benzene
FTNJQNQLEGKTGD-UHFFFAOYSA-N
1,3-benzodioxole
9229
122.12
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning

EINECSNumber : 205-992-0

RTECSNumber : DA5600000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only