Please login to your online account to display your discounted pricing

1,3-Benzodioxole, 99%, ACROS Organics™


Quantity 50g

Manufacturer: thermo fisher scientific  170220500

 View more versions of this product

Catalog No. AC170220500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Physical Form Powder
Formula Weight 122.12
Quantity 50g
Color Yellow to orange to brown
TSCA TSCA
RTECS Number DA5600000
EINECS Number 205-992-0
CAS Max % 100.0
Refractive Index 1.5380 to 1.5400 (20°C, 589nm)
Chemical Name or Material 1, 3-Benzodioxole
Health Hazard 1 Warning
Molecular Formula C7H6O2
Name Note 99%
CAS Min % 98.5
MDL Number MFCD00005818
CAS 274-09-9
Assay Percent Range 99%
Synonym 1, 2-(Methylenedioxy)benzene
Water 0.1% max.
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Flammable liquid and vapor.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1, 3-Benzodioxole
Name Note 99%
CAS 274-09-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C7H6O2
Formula Weight 122.12
MDL Number MFCD00005818
Synonym 1, 2-(Methylenedioxy)benzene
Accessories

Warning

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Flammable liquid and vapor.

SDS & Safety Documents