missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Benzodioxole, 99%, ACROS Organics™

$45.72 - $45.72

Chemical Identifiers

CAS 274-09-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00005818
InChI Key FTNJQNQLEGKTGD-UHFFFAOYSA-N
Synonym 1,2-methylenedioxy benzene, 1,2-methylenedioxybenzene, 1,3-dioxaindan, 1,3-dioxindan, 2h-1,3-benzodioxole, 3,4-methylenedioxybenzene, benzo d 1,3 dioxole, benzodioxole, methylenedioxybenzene, o-methylenedioxy benzene
PubChem CID 9229
ChEBI CHEBI:38732
IUPAC Name 1,3-benzodioxole
SMILES C1OC2=CC=CC=C2O1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC170220500
SDS View Documents
Thermo Fisher Scientific
170220500
50g
Each for $45.72
 
AC170222500
SDS View Documents
Thermo Fisher Scientific
AC170222500
250g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC170220010
SDS View Documents
Thermo Fisher Scientific
AC170220010
1kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

274-09-9
122.12
FTNJQNQLEGKTGD-UHFFFAOYSA-N
9229
1,3-benzodioxole
C7H6O2
MFCD00005818
1,2-methylenedioxy benzene, 1,2-methylenedioxybenzene, 1,3-dioxaindan, 1,3-dioxindan, 2h-1,3-benzodioxole, 3,4-methylenedioxybenzene, benzo d 1,3 dioxole, benzodioxole, methylenedioxybenzene, o-methylenedioxy benzene
CHEBI:38732
C1OC2=CC=CC=C2O1

Specifications

1.0600g/mL
Authentic
1.5380 to 1.5400
0.1% max.
Brown to Yellow
50g
98.5
98.5% min. (GC)
MFCD00005818
1,2-methylenedioxy benzene, 1,2-methylenedioxybenzene, 1,3-dioxaindan, 1,3-dioxindan, 2h-1,3-benzodioxole, 3,4-methylenedioxybenzene, benzo d 1,3 dioxole, benzodioxole, methylenedioxybenzene, o-methylenedioxy benzene
FTNJQNQLEGKTGD-UHFFFAOYSA-N
1,3-benzodioxole
9229
122.12
99%
55°C
Glass bottle
1.06
172.0°C to 173.0°C
-18.0°C
274-09-9
100.0
C7H6O2
19,2
Solubility in water: 0.2% (25 c). Other solubilities: miscible with most common organic solvents
C1OC2=CC=CC=C2O1
122.12
CHEBI:38732
Powder
1, 3-Benzodioxole, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning

EINECSNumber : 205-992-0

RTECSNumber : DA5600000

TSCA : TSCA

SDS