Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Benzodioxole, 99%

$49.11 - $825.70

Chemical Identifiers

Chemical Name or Material 1,3-Benzodioxole
CAS 274-09-9
Molecular Formula C7H6O2
Formula Weight 122.12
MDL Number MFCD00005818
Beilstein 115506
Synonym 1,2-(Methylenedioxy)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA141920B

 
alfa aesar™
A141920B
1000g Each for $825.70

AAA1419218

 
alfa aesar™
A1419218
50g Each for $49.11

AAA1419230

 
alfa aesar™
A1419230
250g Each for $221.50
Description & Specifications

Specifications

Formula Weight 122.12
Assay 99%
Quantity 1000g
Beilstein 115506
Health Hazard 1 Warning
CAS 274-09-9
RTECS Number DA5600000
Synonym 1,2-(Methylenedioxy)benzene
Health Hazard 2 GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Chemical Name or Material 1,3-Benzodioxole
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 55°C (131°F)
Odor Aromatic
Molecular Formula C7H6O2
Melting Point −18°C
MDL Number MFCD00005818
TSCA TSCA
Boiling Point 172° to 173°C

Applications
1,3-Benzodioxole is useful in gemological stimulant detection. It is also used as a precursor for perfumes, photo initiators, agrochemicals and pharmaceuticals.

Solubility
Soluble in water (2g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.