Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,3-Benzenedithiol, 97%

$129.58 - $477.65

Chemical Identifiers

Chemical Name or Material 1,3-Benzenedithiol
CAS 626-04-0
Molecular Formula C6H6S2
Formula Weight 142.24
MDL Number MFCD00004842
Beilstein 1634412
Synonym 1,3-Dimercaptobenzene; Dithioresorcinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1943903

  SDS
alfa aesar™
L1943903
1g Each for $129.58

AAL1943906

  SDS
alfa aesar™
L1943906
5g Each for $477.65
Description & Specifications

Specifications

Formula Weight 142.24
CAS 626-04-0
Chemical Name or Material 1,3-Benzenedithiol
Assay 97%
Molecular Formula C6H6S2
MDL Number MFCD00004842
Synonym 1,3-Dimercaptobenzene; Dithioresorcinol
Health Hazard 1 Danger
Boiling Point 244°C
Quantity 1g
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1634412
Flash Point >112°C (234°F)
Odor Stench
Recommended Storage 2° to 8°C

Applications
1,3-Benzenedithiol is used to improve power conversion efficiency in cadmium selenide nanorods/poly(3-hexylthiophene) hybrid solar cells. It acts as an intermediate as well as involved in the preparation of 1,3-bis(-styrylthio)-benzol.

Solubility
Miscible with acetonitrile.

Notes
Incompatible with strong bases.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

2° to 8°C