missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Benzenedisulfonyl chloride, 98%, ACROS Organics™
$266.98 - $266.98
Chemical Identifiers
CAS | 585-47-7 |
---|---|
Molecular Formula | C6H4Cl2O4S2 |
Molecular Weight (g/mol) | 275.13 |
MDL Number | MFCD00041879 |
InChI Key | ALIQZUMMPOYCIS-UHFFFAOYSA-N |
Synonym | 1,3-benzenedisulfonyl chloride, 1,3-benzenedisulfonyl dichloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, acmc-209m5a, benzene-1,3-di sulphonyl chloride, benzene-1,3-disulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride |
PubChem CID | 96240 |
IUPAC Name | benzene-1,3-disulfonyl chloride |
SMILES | C1=CC(=CC(=C1)S(=O)(=O)Cl)S(=O)(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC401810250
|
Thermo Fisher Scientific
401810250 |
25g | Glass bottle |
Each for $266.98
|
|
Chemical Identifiers
585-47-7 | |
275.13 | |
ALIQZUMMPOYCIS-UHFFFAOYSA-N | |
96240 | |
C1=CC(=CC(=C1)S(=O)(=O)Cl)S(=O)(=O)Cl |
C6H4Cl2O4S2 | |
MFCD00041879 | |
1,3-benzenedisulfonyl chloride, 1,3-benzenedisulfonyl dichloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, acmc-209m5a, benzene-1,3-di sulphonyl chloride, benzene-1,3-disulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride | |
benzene-1,3-disulfonyl chloride |
Specifications
Authentic | |
White to Beige | |
25g | |
97.5 | |
98% | |
C6H4(SO2Cl)2 | |
1,3-benzenedisulfonyl chloride, 1,3-benzenedisulfonyl dichloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, acmc-209m5a, benzene-1,3-di sulphonyl chloride, benzene-1,3-disulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride | |
ALIQZUMMPOYCIS-UHFFFAOYSA-N | |
benzene-1,3-disulfonyl chloride | |
96240 | |
Crystalline Powder | |
1, 3-Benzenedisulfonyl chloride, 98% |
Glass bottle | |
56°C to 58°C | |
585-47-7 | |
100.0 | |
C6H4Cl2O4S2 | |
MFCD00041879 | |
Solubility in water: reacts | |
C1=CC(=CC(=C1)S(=O)(=O)Cl)S(=O)(=O)Cl | |
275.13 | |
275.13 | |
98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 209-556-0
TSCA : TSCA