missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3-Benzenedisulfonamide, TCI America™

$247.50 - $742.52

Chemical Identifiers

CAS 3701-01-7
Molecular Formula C6H8N2O4S2
Molecular Weight (g/mol) 236.26
MDL Number MFCD00025383
InChI Key HUYYFHGIHVULSU-UHFFFAOYSA-N
PubChem CID 3090044
ChEBI CHEBI:35085
IUPAC Name benzene-1,3-disulfonamide
SMILES C1=CC(=CC(=C1)S(=O)(=O)N)S(=O)(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B49171G View Documents TCI America
B49171G
1g Each for $742.52
 
B4917200MG View Documents TCI America
B4917200MG
200mg Each for $247.50
 
Specifications

Chemical Identifiers

3701-01-7
236.26
HUYYFHGIHVULSU-UHFFFAOYSA-N
CHEBI:35085
C1=CC(=CC(=C1)S(=O)(=O)N)S(=O)(=O)N
C6H8N2O4S2
MFCD00025383
3090044
benzene-1,3-disulfonamide

Specifications

White
1g
C6H8N2O4S2
HUYYFHGIHVULSU-UHFFFAOYSA-N
benzene-1,3-disulfonamide
3090044
236.26
≥98.0% (HPLC,N)
235°C
3701-01-7
MFCD00025383
C1=CC(=CC(=C1)S(=O)(=O)N)S(=O)(=O)N
236.26
CHEBI:35085
Crystal-Powder at 20°C
1,3-Benzenedisulfonamide
Safety and Handling

Safety and Handling

TSCA : No