missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,3-Benzenedimethanol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 188200020
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 3-Benzenedimethanol | |
626-18-6 | |
100.0 | |
Authentic | |
Glass bottle | |
C6H4(CH2OH)2 | |
2g | |
1,3-benzenedimethanol, 3-hydroxymethyl phenyl methanol, 1,3-phenylenedimethanol, 1,3-dihydroxymethylbenzene, m-xylene-alpha,alpha'-diol, m-xylene glycol, m-xylylene glycol, 3-hydroxymethyl phenyl methan-1-ol, m-xylylenalkohol, m-benzenedimethanol | |
C1=CC(=CC(=C1)CO)CO | |
138.17 | |
138.17 |
98% | |
97.5 | |
154.0°C to 159.0°C (13.0mmHg) | |
97.5% min. (GC) | |
C8H10O2 | |
MFCD00004649 | |
06, 914 | |
YWMLORGQOFONNT-UHFFFAOYSA-N | |
[3-(hydroxymethyl)phenyl]methanol | |
69374 | |
98% |
Chemical Identifiers
626-18-6 | |
138.17 | |
YWMLORGQOFONNT-UHFFFAOYSA-N | |
69374 | |
C1=CC(=CC(=C1)CO)CO |
C8H10O2 | |
MFCD00004649 | |
1,3-benzenedimethanol, 3-hydroxymethyl phenyl methanol, 1,3-phenylenedimethanol, 1,3-dihydroxymethylbenzene, m-xylene-alpha,alpha'-diol, m-xylene glycol, m-xylylene glycol, 3-hydroxymethyl phenyl methan-1-ol, m-xylylenalkohol, m-benzenedimethanol | |
[3-(hydroxymethyl)phenyl]methanol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 210-934-2
TSCA : TSCA
RUO â Research Use Only