Please login to your online account to display your discounted pricing

1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one
CAS 175137-02-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10ClNOS
Formula Weight 251.74
MDL Number MFCD00068162
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

GK00604CB

 
fisher scientific company
GK00604CB
Amber glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GK00604DA

 
fisher scientific company
GK00604DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 251.74
Quantity 250mg
Assay 97%
CAS Max % 100.0
Packaging Amber glass bottle
MDL Number MFCD00068162
Chemical Name or Material 1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one
Assay Percent Range 97%
Molecular Formula C12H10ClNOS
CAS 175137-02-7
CAS Min % 96.5