Please login to your online account to display your discounted pricing

1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one, 97%, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  GK00604DA

 View more versions of this product

Catalog No. GK00604DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 251.74
Quantity 1g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one
Packaging Amber glass bottle
Assay Percent Range 97%
MDL Number MFCD00068162
Molecular Formula C12H10ClNOS
CAS 175137-02-7
CAS Min % 96.5

Specifications

Chemical Name or Material 1-[3-Amino-5-(4-chlorophenyl)-2-thienyl]ethan-1-one
CAS 175137-02-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10ClNOS
Formula Weight 251.74
MDL Number MFCD00068162