missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(3,6-Dibromo-9H-carbazol-9-yl)-3-(dimethylamino)propan-2-ol, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company BTB12241DA

 View more versions of this product

Catalog No. BTB12241DA



Description

Description

Specifications

Specifications

1-(3, 6-Dibromo-9H-carbazol-9-yl)-3-(dimethylamino)propan-2-ol
1g
96.5
97%
MFCD00218393
XUBJEDZHBUPBKL-UHFFFAOYSA-N
1-(3,6-dibromocarbazol-9-yl)-3-(dimethylamino)propan-2-ol
2775510
426.15
Amber Glass Bottle
253449-04-6
100.0
C17H18Br2N2O
1-3,6-bis bromanyl carbazol-9-yl-3-dimethylamino propan-2-ol, 1-3,6-dibromo-9-carbazolyl-3-dimethylamino-2-propanol, 1-3,6-dibromo-9h-carbazol-9-yl-3-dimethylamino, 1-3,6-dibromo-9h-carbazol-9-yl-3-dimethylamino propan-2-ol, 1-3,6-dibromo-9h-carbazol-9-yl-3-dimethylamino propan-2ol, 1-3,6-dibromo-carbazol-9-yl-3-dimethylamino-propan-2-ol, 1-3,6-dibromocarbazol-9-yl-3-dimethylamino propan-2-ol, 3,6-dibromo-?-dimethylamino methyl-9h-cabazole-9-ethanol, maybridge1_002006, wiskostatin
CN(C)CC(CN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br)O
426.152
CHEBI:78012
97%

Chemical Identifiers

253449-04-6
426.152
XUBJEDZHBUPBKL-UHFFFAOYSA-N
2775510
1-(3,6-dibromocarbazol-9-yl)-3-(dimethylamino)propan-2-ol
C17H18Br2N2O
MFCD00218393
1-3,6-bis bromanyl carbazol-9-yl-3-dimethylamino propan-2-ol, 1-3,6-dibromo-9-carbazolyl-3-dimethylamino-2-propanol, 1-3,6-dibromo-9h-carbazol-9-yl-3-dimethylamino, 1-3,6-dibromo-9h-carbazol-9-yl-3-dimethylamino propan-2-ol, 1-3,6-dibromo-9h-carbazol-9-yl-3-dimethylamino propan-2ol, 1-3,6-dibromo-carbazol-9-yl-3-dimethylamino-propan-2-ol, 1-3,6-dibromocarbazol-9-yl-3-dimethylamino propan-2-ol, 3,6-dibromo-?-dimethylamino methyl-9h-cabazole-9-ethanol, maybridge1_002006, wiskostatin
CHEBI:78012
CN(C)CC(CN1C2=C(C=C(C=C2)Br)C3=C1C=CC(=C3)Br)O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents