missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,6,8-Tetrabromocarbazole, TCI America™

Manufacturer:  TCI America T29321G

 View more versions of this product

Catalog No. T29321G



Specifications

Specifications

1,3,6,8-Tetrabromocarbazole
229°C
55119-09-0
MFCD00218272
C1=C(C=C2C3=CC(=CC(=C3NC2=C1Br)Br)Br)Br
482.795
482.80
≥95.0% (GC)
White-Yellow
1g
C12H5Br4N
FNHISDQCWYSMTO-UHFFFAOYSA-N
1,3,6,8-tetrabromo-9H-carbazole
258744
Crystal-Powder at 20°C

Chemical Identifiers

55119-09-0
482.795
FNHISDQCWYSMTO-UHFFFAOYSA-N
1,3,6,8-tetrabromo-9H-carbazole
C12H5Br4N
MFCD00218272
258744
C1=C(C=C2C3=CC(=CC(=C3NC2=C1Br)Br)Br)Br
Safety and Handling

Safety and Handling

TSCA : No

Documents